Skip to Content
Merck
All Photos(1)

Documents

567715

Sigma-Aldrich

N-SMase Inhibitor, GW4869

The N-SMase Inhibitor, GW4869, also referenced under CAS 6823-69-4, controls the biological activity of N-SMase. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

N-SMase Inhibitor, GW4869, Sphingomyelinase, Neutral, Inhibitor GW4869, GW69A, GW554869A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H28N6O2 · 2HCl
CAS Number:
Molecular Weight:
577.50
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

pale yellow

solubility

DMSO: 200 μg/mL

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C30H28N6O2.2ClH/c37-27(35-25-11-7-23(8-12-25)29-31-17-18-32-29)15-5-21-1-2-22(4-3-21)6-16-28(38)36-26-13-9-24(10-14-26)30-33-19-20-34-30;;/h1-16H,17-20H2,(H,31,32)(H,33,34)(H,35,37)(H,36,38);2*1H/b15-5+,16-6+;;

InChI key

NSFKAZDTKIKLKT-CLEIDKRQSA-N

General description

A cell permeable, symmetrical dihydroimidazolo-amide that acts as a potent, specific, and non-competitive inhibitor of neutral sphingomyelinase (N-SMase) (IC50 = 1 µM, rat brain N-SMase). Reported to inhibit tumor necrosis factor α (TNF-α) induced sphingomyelin hydrolysis (100% inhibition at 20 µM) and TNF-α-induced cell death in MCF7 cells. Does not interfere with other TNF-α-mediated signaling events, such as de novo ceramide synthesis and NF-κB activation and nuclear translocation. Exhibits no effect on cellular glutathione levels that are normally reduced in response to TNF-α. Does not inhibit acid sphingomyelinase (A-SMase) or bacterial phosphatidylcholine-specific phospholipase C (PC-PLC). Shown to weakly inhibit bovine protein phosphatase 2A (PP2A) and human lyso-PAF PLC, an enzyme that hydrolyzes sphingomyelin in vitro.
A cell-permeable, symmetrical dihydroimidazolo-amide compound that acts as a potent, specific, non-competitive inhibitor of N-SMase (neutral sphingomyelinase) [IC50 = ~ 1 µM, rat brain; Km for sphingomyelin ~13 µM]. Does not inhibit human A-SMase (acid sphingomyelinase) even at 150 µM. Weakly inhibits the activities of bovine protein phosphatase 2A and mammalian lyso-PAF PLC, while no inhibition is observed for bacterial phosphatidylcholine-specific PLC. Reported to offer complete protection against TNF-α or diamine-induced cell death in MCF7 breast cancer cells at 20 µM. Does not modify the intracellular glutathione levels or interfere with TNF-α or diamine-mediated signaling effects.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
N-Smase rat brain
Product does not compete with ATP.
Reversible: no
Target IC50: 1 µM rat brain N-SMase (neutral sphingomyelinase); Km for sphingomyelin ~13 µM

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Marchesini, N., et al. 2003. J. Biol. Chem. In press.
Luberto, C., et al. 2002. J. Biol. Chem.277, 41128
Okamoto, Y., et al. 2002. FEBS Lett.530, 140.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huimin Huang et al.
Journal of orthopaedic surgery and research, 18(1), 687-687 (2023-09-15)
Diabetic foot ulcers (DFU) are a serious complication of diabetes that lead to significant morbidity and mortality. Recent studies reported that exosomes secreted by human adipose tissue-derived mesenchymal stem cells (ADSCs) might alleviate DFU development. However, the molecular mechanism of
Christian M Garrido et al.
Scientific reports, 10(1), 9120-9120 (2020-06-06)
K-Ras must interact primarily with the plasma membrane (PM) for its biological activity. Therefore, disrupting K-Ras PM interaction is a tractable approach to block oncogenic K-Ras activity. Here, we found that avicin G, a family of natural plant-derived triterpenoid saponins
Cuiqi Zhou et al.
The Journal of clinical endocrinology and metabolism, 107(2), 379-397 (2021-09-02)
The identification and biological actions of pituitary-derived exosomes remain elusive. This work aimed to validate production of exosomes derived from human and rat pituitary and elucidate their actions. Isolated extracellular vesicles (EVs) were analyzed by Nanoparticle Tracking Analysis (NTA) and
Lara Ottaviani et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 30(6), 2257-2273 (2022-03-13)
As mediators of intercellular communication, extracellular vesicles containing molecular cargo, such as microRNAs, are secreted by cells and taken up by recipient cells to influence their cellular phenotype and function. Here we report that cardiac stress-induced differential microRNA content, with
Jian Shi et al.
Cell reports, 33(1), 108225-108225 (2020-10-08)
Endogenous PIEZO1 channels of native endothelium lack the hallmark inactivation often seen when these channels are overexpressed in cell lines. Because prior work showed that the force of shear stress activates sphingomyelinase in endothelium, we considered if sphingomyelinase is relevant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service