Skip to Content
Merck
All Photos(1)

Key Documents

554716

Sigma-Aldrich

REV-ERBα Agonist, GSK4112

The REV-ERBα Agonist, GSK4112 controls the biological activity of REV-ERBα. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

REV-ERBα Agonist, GSK4112, GSK4112

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€108.00

€108.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€108.00

About This Item

Empirical Formula (Hill Notation):
C18H21ClN2O4S
CAS Number:
Molecular Weight:
396.89
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€108.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark beige

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[s]1c(ccc1CN(Cc2ccc(cc2)Cl)CC(=O)OC(C)(C)C)[N+](=O)[O-]

InChI key

WYSLOKHVFKLWOU-UHFFFAOYSA-N

General description

A selective, cell-permeable ligand (EC50 = 250 nM) that acts as a REV-ERBα agonist using a FRET assay. It enhances the recruitment of NCoR peptide to REV-ERBα by up to 70%, and displays no activity towards LRH1, SF1, FXR or ROR, and LXR. Furthermore, it inhibits the promoter activity of Pai1, a known target for REV-ERBα by 30%. This ligand exhibits acute suppression of Bmal1 transcription, and is capable of resetting the molecular rhythm of REV-ERBα-mediated PER2 expression in both Rat-1 lung fibroblasts and on ectopic lung tissues in Luciferase reporter assays. In addition, it is shown to repress the expression of gluconeogenic genes in liver cells and reduce glucose output in primary hepatocytes.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Grant, D., et al. 2010, ACS Chem. Biol.5, 925.
Meng, Q., et al. 2008. Journ. Cell. Sci121, 3629.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service