Skip to Content
Merck
All Photos(1)

Key Documents

5.38763

Sigma-Aldrich

CD38 Inhibitor

≥98% (HPLC), solid, CD38 inhibitor, Calbiochem®

Synonym(s):

CD38 Inhibitor, Compound 78c, CD38i_78c, 78c, Cluster of Differentiation 38 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€231.00

€231.00


Estimated to ship on28 May 2025



Select a Size

Change View
10 MG
€231.00

About This Item

Empirical Formula (Hill Notation):
C22H27N3O3S
CAS Number:
Molecular Weight:
413.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€231.00


Estimated to ship on28 May 2025


Product Name

CD38 Inhibitor,

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light brown to brown
white to light brown

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

[s]1cncc1c2cc3c(cc2)N(C(=O)C=C3N[C@@H]4CC[C@H](CC4)OCCOC)C

InChI

1S/C22H27N3O3S/c1-25-20-8-3-15(21-13-23-14-29-21)11-18(20)19(12-22(25)26)24-16-4-6-17(7-5-16)28-10-9-27-2/h3,8,11-14,16-17,24H,4-7,9-10H2,1-2H3/t16-,17-

InChI key

VJQALSOBHVEJQM-QAQDUYKDSA-N

General description

A cell-permeable, orally bioavailable, non-toxic thiazolyl-quinolinone derived compound that acts as a potent, selective and reversible inhibitor of CD38 (IC50 = 7.3 & 1.9 nM for human & mouse CD38, respectively; Ki = 0.3 nM for human wt-CD38). Shown to bind to the catalytic active site. Displays attractive PK properties in mice (p.o. 30 mg/kg: t1/2 = 3.5 h, Cmax = 10.8 µg/ml; i.v. 2 mg/kg: t1/2 = 5.3 h, Cmax = 2.98 µg/ml, Cl = 21 ml/min/kg, Vdss = 1.3 L/kg). Rapidly elevates NAD levels by several folds in both liver and gastrocnemius tissue in a diet induced obese mouse (30 mg/kg, p.o.).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CD38
Reversible: yes
Target IC50: 7.3 & 1.9 nM for human & mouse CD38, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Haffner, C.D., et al. 2015. J. Med. Chem.58, 3548.
Tarrago, M.G., et al. 2018 Cell. Metab.27, 1081.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service