Skip to Content
Merck
All Photos(2)

Key Documents

5.38299

Sigma-Aldrich

M2I-1

Synonym(s):

M2I-1, 5-(4-(Diisobutylamino)-3-nitrobenzylidene)-2-thioxodihydro-4,6(1H,5H)-pyrimidinedione, 5-(4-(Diisobutylamino)-3-nitrobenzylidene)-2-thioxodihydropyrimidine-4,6(1H,5H)-dione, Mad 2 Inhibitor- 1, 5-(4-(Diisobutylamino)-3-nitrobenzylidene)-2-thioxodihydro-4,6(1H,5H)-pyrimidinedione, 5-(4-(Diisobutylamino)-3-nitrobenzylidene)-2-thioxodihydropyrimidine-4,6(1H,5H)-dione, Mad2Inhibitor-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N4O4S
Molecular Weight:
404.48
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

orange

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable nitrobenzylidene-thiobarbiturate compound that directly targets the hydrophobic core of Mad2 and selectively disrupts Mad2-Cdc20 complex formation. Shown to dose-dependently inhibit the interaction of Mad2 to Mad2-binding-peptide (Mbp1; IC50 ~ 10.0 µM & Kd = 200 nM). Suggested to covalently modify Mad2-Cys149 via a reversible Michael adduct formation. Significantly reduces the mitotic duration in paclitaxel (0.5 µM; Cat. No. 580555)-treated HeLa cells (25 µM of M2I-1) and impairs mitotic spindle assembly checkpoint (SAC) function.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Mad2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Kastl, J., et al. 2015. ACS Chem. Biol.10, 1661.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service