Skip to Content
Merck
All Photos(1)

Key Documents

5.32978

Sigma-Aldrich

PLD2 Inhibitor, ML395

Synonym(s):

PLD2 Inhibitor, ML395, Phosppolipase D2 inhibitor, VU0468809, ML-395

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€192.00

€192.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€192.00

About This Item

Empirical Formula (Hill Notation):
C26H29N5O2
CAS Number:
Molecular Weight:
443.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€192.00


Estimated to ship on19 May 2025


Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

solid

potency

360 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell permeable, triazaspirone derivative that acts as a highly potent, selective, and direct allosteric inhibitor of phospholipase D2 (PLD2; IC50 = 360 nM in exogenous biochemical assay). Exhibits >80-fold selectivity over phospholipase D1 (PLD1; IC50 = 30 µM). Shown to permeate the blood-brain barrier. Protects A549 cells from multiple strains of influenza virus when cells were pre-treated with this compound. Exhibits excellent DMPK profile (hepatic microsomal clearance = 82.1 ml/min/kg in Sprague-Dawley rats) and conforms to Lipinski′s rule and has favorable lipophilicity. Also displays favorable cytochrome P450 profile (CYP3A4 IC50 = 3.9 µM, CYP2D6 IC50 =16.4 µM, CYP1A2 IC50>30 µM, and CYP2C9 IC50>30 µM).
ML395 is a cell permeable, highly potent, selective, and direct allosteric inhibitor of phospholipase D2 (PLD2; IC₅₀ = 360 nM) that exhibits >80-fold selectivity over PLD1.

Biochem/physiol Actions

Primary Target
PLD2
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C).

Other Notes

O′Reilly, M.C., et al. 2014. ChemMedChem9, 2633.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service