5.29566
Silicone tubing for booster Ø 6,4 mm
for use with Dilucult™, pkg of 5 tubes
Synonym(s):
Dilutor Tubing, Hose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
silicone tubing
packaging
pkg of 5 tubes
manufacturer/tradename
DiluCult™
application(s)
food and beverages
compatibility
for use with Dilucult™
General description
Set of 5 dispensing tubings (silicone tubings + tygon tubings + "Y" connectors) - inner ø 6.4 mm
Application
For use with DiluCult™ and DiluCult™ 2 gravimetric dilutor instrument
Legal Information
DILUCULT is a trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service