Skip to Content
Merck
All Photos(1)

Documents

528110-M

Millipore

PIH

A cell-permeable, non-toxic tridendate iron (Fe3+) chelator of the aroyl hydrazone class.

Synonym(s):

PIH, pyridoxal isonicotinoyl hydrazone, Iron Chelator III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N4O3
CAS Number:
Molecular Weight:
286.29
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥99% (HPLC)

form

solid

reaction suitability

reagent type: chelator

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL
slighlty acidic medium: soluble

storage temp.

2-8°C

InChI

1S/C14H14N4O3/c1-9-13(20)12(11(8-19)6-16-9)7-17-18-14(21)10-2-4-15-5-3-10/h2-7,17,19H,8H2,1H3,(H,18,21)/b12-7+

InChI key

HOYFGBFEIXEDEL-KPKJPENVSA-N

General description

A cell-permeable, non-toxic tridendate iron (Fe3+) chelator of the aroyl hydrazone class. Even at low concentrations, highly effective in mobilizing Fe3+ from cells and preventing its uptake from transferrin, even at low concentrations. Exhibits similar specificity and binding affinity as Deferoxamine Mesylate (Cat. No. 252750) but has greater access to mitochondrial iron. Also useful for the management of iron overload disease in experimental models. Inhibits the induction of heme containing indoleamine 2,3-dioxygenase activity.
A cell-permeable, non-toxic, iron (Fe3+) chelator of the aroyl hydrazone class that exhibits high chelation efficacy. Highly effective in mobilizing Fe3+ from cells and preventing its uptake from transferrin, even at low concentrations. Exhibits similar specificity and binding affinity as Desferrioxamine (Cat. No. 252750) but has greater access to mitochondrial iron. Prevents iron-mediated oxyradical formation and minimizes tissue damage. Reported to useful in studies of the management of iron overload disease. Inhibits the induction of heme-containing indoleamine 2,3-dioxygenase activity.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 6 months at -20°C. Acidic stock solutions are stable for up to 2 months at -20°C.

Other Notes

Buss, J.L., et al. 2002. Biochem. Pharmacol.64, 1689.
Thomas, S.R., et al. 2001. J. Immunol.166, 6332.
Hermes-Linda, M., et al. 2000. Biochim. Biophys. Acta1523, 154.
Blaha, K., et al. 1998. Blood11, 4368.
Richardson, D.R., and Ponka, P. 1998. J. Lab. Clin. Med.131, 306.
Richardson, D.R.,et al. 1995. Blood86, 4295.
Richardson, D.R., et al. 1994. J. Lab. Clin. Med.124, 660.
Ponka, P., et al. 1988. Biochim. Biophys. Acta967, 122.
Ponka, P., et al. 1984. Biochim. Biophys. Acta802, 477.
Ponka, P., et al. 1979. FEBS Lett.97, 317.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service