Skip to Content
Merck
All Photos(1)

Documents

527455

Sigma-Aldrich

PKR Inhibitor, Negative Control

The PKR Inhibitor, Negative Control, also referenced under CAS 852547-30-9, controls the biological activity of PKR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PKR Inhibitor, Negative Control, RNA-Dependent Protein Kinase Inhibitor, Negative Control, 5-Chloro-3-(3,5-dichloro-4-hydroxybenzylidene)-1,3-dihydro-indol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8Cl3NO2
CAS Number:
Molecular Weight:
340.59
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL
ethanol: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C15H8Cl3NO2/c16-8-1-2-13-9(6-8)10(15(21)19-13)3-7-4-11(17)14(20)12(18)5-7/h1-6,20H,(H,19,21)

InChI key

ZJFMARHFPUZEKI-UHFFFAOYSA-N

General description

An oxindole compound that serves as a negative control for PKR Inhibitor (Cat. No. 527450). Shown to be inactive in inhibiting RNA-induced PKR autophosphorylation (IC50 >100 µM) or in rescuing PKR-dependent translation block.
An oxindole compound that serves as a negative control for PKR Inhibitor (Cat. No. 527450). Shown to be inactive in the inhibition of RNA-induced PKR autophosphorylation (IC50 >100 µM). Exhibits no ability to rescue the translational block imposed by human PKR.

Biochem/physiol Actions

Cell permeable: no
Primary Target
A negative control for PKR Inhibitor
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Jammi, N.V., et al. 2003. Biochem. Biophys. Res. Commun.308, 50.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanket S Ponia et al.
Cell reports, 37(4), 109888-109888 (2021-10-28)
Dysregulated inflammation dominated by chemokine expression is a key feature of disease following infection with the globally important human pathogens Zika virus (ZIKV) and dengue virus, but a mechanistic understanding of how pro-inflammatory responses are initiated is lacking. Mitophagy is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service