Skip to Content
Merck
All Photos(1)

Documents

5250-1BOTTLE

Millipore

Acid Buffer pH 2.2

new

ready-to-use, glass bottle of 450 mL, selective for bacteria: Legionella spp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217

Agency

ISO 11731

form

liquid

feature

ready-to-use

packaging

glass bottle of 450 mL

technique(s)

microbiological culture: suitable

pH

2.2

application(s)

food and beverages
water monitoring

storage temp.

2-25°C

suitability

selective for bacteria: Legionella spp

General description

Acid Buffer pH 2.2 is designed as a critical media component for Legionella spp. studies. Legionella a gram-negative, rod-shaped bacteria, are commonly found in various freshwater biotopes, often associated with protozoans such as amoebae, where they can grow intracellularly.
The presence of acid in the growth medium inhibits colony formation of other contaminating bacteria from the test sample.

Application

Acid Buffer with pH 2.2 is used for selective growth of Legionella in water samples from food and beverage industry.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service