Skip to Content
Merck
All Photos(1)

Key Documents

513028

Sigma-Aldrich

PD 166285

A cell-permeable, orally bioavailable, ATP-competitive, broad-spectrum tyrosine kinase inhibitor that suppresses angiogenesis both in vitro and in vivo.

Synonym(s):

PD 166285, PD166285, PD0166285, 6-(2,6-Dichlorophenyl)-2-(4-(2-(diethylaminoethoxy)-phenylamino)-8-methyl-8H-pyrido[2,3-d]pyrimidin-7-one, 2HCl, PDGFR Tyrosine Kinase Inhibitor XIX, Wee1 Inhibitor IV, PDGFR Tyrosine Kinase Inhibitor XIX, Wee1 Inhibitor IV, 6-(2,6-Dichlorophenyl)-2-(4-(2-(diethylaminoethoxy)-phenylamino)-8-methyl-8H-pyrido[2,3-d]pyrimidin-7-one, 2HCl, PD166285, PD0166285

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H27Cl2N5O2 · 2HCl
CAS Number:
Molecular Weight:
585.35
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C26H27Cl2N5O2.2ClH/c1-4-33(5-2)13-14-35-19-11-9-18(10-12-19)30-26-29-16-17-15-20(25(34)32(3)24(17)31-26)23-21(27)7-6-8-22(23)28;;/h6-12,15-16H,4-5,13-14H2,1-3H3,(H,29,30,31);2*1H

InChI key

NADLBPWBFGTESN-UHFFFAOYSA-N

General description

A cell-permeable, orally bioavailable, ATP-competitive, broad-spectrum tyrosine kinase inhibitor (IC50 against against c-Src, Wee1, FGFR-1, Myt1, EGFR, and PDGFRβ = 8.4, 24, 39.3, 72, 87.5 and 98.3 nM, respectively) that suppresses angiogenesis both in vitro (max inhibition dose at 100 nM in HUVEC microcapillary formation assays) and in vivo (max inhibition achieved via 5 mg/kg p.o. in murine Matrigel plug angiogenesis assays), while exhibiting much reduced potency against Chk1, MAPK, and PKC (IC50 = 3.4, 5, and 22.7 µM, respectively) and little activity toward IRTK and Cdk4/D1 even at concentrations as high as 50 µM. Shown to effectively block PDGF-, EGF-, and bFGF-stimulated receptor phosphorylations (IC50 = 6.5, 1600, and 97.3 nM, respectively) and other cellular responses in rat aortic smooth muscle cells. Inhibition of cellular Wee1 activity by 500 nM PD 166285 in combination with 50 ng/ml nocodazole (Cat. No. 487928) treatment is also reported to result in a blockage of radiation-induced Cdc2 phosphorylation on Tyr15 and Thr14 in 7 human cancer cells and specifically demonstrated to sensatize PA-1 cultures to radiation-induced cell death in a p53-dependent manner.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wang, Y., et al. 2001. Cancer Res.61, 8211.
Dimitroff, C.J., et al. 1999. Invest. New Drugs17, 121.
Roginskaya, V., et al. 1999. Leukemia13, 855.
Panek, R.L., et al. 1997. J. Pharmacol. Exp. Ther.283, 1433.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Lockhead et al.
Cell reports, 32(2), 107901-107901 (2020-07-16)
Protein synthesis inhibitors (e.g., cycloheximide) block mitotic entry, suggesting that cell cycle progression requires protein synthesis until right before mitosis. However, cycloheximide is also known to activate p38 mitogen-activated protein kinase (MAPK), which can delay mitotic entry through a G2/M

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service