5.08941
GPR40 Agonist III
Synonym(s):
GPR40 Agonist III, FFA1 Agonist III, Free Fatty Acid Receptor 1 Agonist III
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥98% (HPLC)
Quality Level
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white
solubility
DMSO: 100 mg/mL
storage temp.
2-8°C
General description
A cell-permeable, orally bioavailable, non-toxic, alkyne derivative that acts as a highly potent, and selective agonist of free fatty acid receptor 1 (FFA1; GPR40; EC50 = 20 nM in FFA1-transfected HEK-293 cells). Exhibits lower lipophilicity and excellent metabolic stability. Demonstrates over 100-fold greater selectivity over GPR43, GPR41, GPR120, PPARγ and 55 other receptors, transporters, and enzymes. Improves glucose tolerance in type 2 diabetic mice (~50 mg/kg) with efficacy comparable to DPPIV inhibitor, sitagliptin (~10 mg/kg). Displays favorable pharmacokinetic properties with t 1/2= 17 min and Cmax of 5071 (ng/ml).
A cell-permeable, orally bioavailable, non-toxic, alkyne derivative that acts as a highly potent, and selective agonist of free fatty acid receptor 1 (FFA1; GPR40; EC50 = 20 nM in FFA1-transfected HEK-293 cells). Exhibits lower lipophilicity and excellent metabolic stability. Demonstrates over 100-fold greater selectivity over GPR43, GPR41, GPR120, PPARγ and 55 other receptors, transporters, and enzymes. Improves glucose tolerance in type 2 diabetic mice (~50 mg/kg) with efficacy comparable to DPPIV inhibitor, sitagliptin (~10 mg/kg). Displays favorable pharmacokinetic properties with t 1/2= 17 min and Cmax of 5071 (ng/ml).
Please note that the molecular weight for this compound is batch-specific due to variable water content.
Please note that the molecular weight for this compound is batch-specific due to variable water content.
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Christiansen, E., et al. 2013. J. Med. Chem.56, 982.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service