Skip to Content
Merck
All Photos(1)

Key Documents

5.06416

Sigma-Aldrich

Beclin-1 Activator II, retro-inverso TAT-Beclin-1

Synonym(s):

Beclin-1 Activator II, retro-inverso TAT-Beclin-1, (D)-H₂N-RRRQRRKKRGY-GG-TGFEGDHWIEFTANFVNT-CO₂H, Atg6 Activator II, Beclin-1-GAPR-1 Interaction Blocker II, Vps30 Activator II

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€572.00

€572.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€572.00

About This Item

Empirical Formula (Hill Notation):
C164H251N57O45
Molecular Weight:
3741.10
UNSPSC Code:
12352200
NACRES:
NA.77

€572.00


Estimated to ship on25 May 2025


Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 50 mg/mL
water: 50 mg/mL

storage temp.

2-8°C

General description

Tat-beclin 1, a peptide, is derived from a segment of the autophagy protein beclin 1, which binds to human immunodeficiency virus (HIV)-1 Nef. It serves as a potent inducer of autophagy. It interacts with GAPR-1 (GLIPR2), a recently identified negative regulator of autophagy. Tat-beclin 1 has been found to reduce the accumulation of polyglutamine expansion protein aggregates and the replication of various pathogens (including HIV-1) in vitro. Moreover, it has demonstrated the ability to decrease mortality in mice infected with chikungunya or West Nile virus.[1]

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GAPR-1/GLIPR2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

(D)-H-arg-arg-arg-gln-arg-arg-lys-lys-arg-Gly-tyr~Gly-Gly~thr-Gly-phe-glu-Gly-asp-his-trp-ile-glu-phe-thr-ala-asn-phe-val-asn-thr-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Shoji-Kawata, S., et al. 2013. Nature.494, 201.

Legal Information

Sold under license of US Patent 8,722,628.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a candidate therapeutic autophagy-inducing peptide.
Shoji-Kawata S, et al.
Nature, 494(7436), 201-201 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service