Skip to Content
Merck
All Photos(1)

Key Documents

5.06271

Sigma-Aldrich

Ketanserin Tartrate

Synonym(s):

Ketanserin Tartrate, 5-HT 2A Serotonin Receptor Antagonist, Ketanserin, R 41 468

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€133.00

€133.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
€133.00

About This Item

Empirical Formula (Hill Notation):
C22H22FN3O3 · xC4H6O6
CAS Number:
Molecular Weight:
395.43 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€133.00


Estimated to ship on20 May 2025


Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

solid

potency

1.49 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: soluble

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C(=O)C2CCN(CC2)CCN3C(=O)Nc4c(cccc4)C3=O.OC(C(O)C(=O)O)C(=O)O.O

InChI

1S/C22H22FN3O3.C4H6O6.H2O/c23-17-7-5-15(6-8-17)20(27)16-9-11-25(12-10-16)13-14-26-21(28)18-3-1-2-4-19(18)24-22(26)29;5-1(3(7)8)2(6)4(9)10;/h1-8,16H,9-14H2,(H,24,29);1-2,5-6H,(H,7,8)(H,9,10);1H2

InChI key

KJJRKCWDAHTVRL-UHFFFAOYSA-N

General description

A selective 5-HT2A serotonin receptor antagonist with weak adrenergic receptor blocking properties. Strong affinity for 5-HT2AR (Ki = 1.49 nM) relative to other family members like 5-HT2BR (Ki = 740 nM) and 5-HT2CR (Ki = 59 nM). Often used in conjunction with SB 206553 to distinguish 5-HT2AR and 5-HT2CR activity.

Biochem/physiol Actions

Primary Target
5-HT2A

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Ros, R. et al. 2013. PNAS.110, 348.

Knight, A. R. et al. 2004. Naunyn Schmiedebergs Arch. Pharmacol.370, 114.
Aloyo, V. J. and Harvey, J. A. 2000. Eur. J. Pharmacol.406, 163.
Leysen, J. E. et al. 1981. Life Sci.28, 1015.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service