Skip to Content
Merck
All Photos(1)

Documents

506158

Sigma-Aldrich

p38 MAP Kinase Inhibitor VII, SD-169

The p38 MAP Kinase Inhibitor VII, SD-169, also referenced under CAS 1670-87-7, controls the biological activity of p38 MAP Kinase.

Synonym(s):

p38 MAP Kinase Inhibitor VII, SD-169, Indole-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2O
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: 1 mg/mL
DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C9H8N2O/c10-9(12)7-1-2-8-6(5-7)3-4-11-8/h1-5,11H,(H2,10,12)

InChI key

GQMYQEAXTITUAE-UHFFFAOYSA-N

General description

A cell-permeable indole compound that acts as a potent, ATP-competitive, and isozyme-selective p38α MAP kinase inhibitor (IC50 = 3.2 nM). It inhibits p38β only at much higher concentrations (IC50 = 122 nM) and exhibits little activity against p38γ, ERK2, JNK-1, or MAPKAPK-2 even at concentrations as high as 50 µM. When administered to NOD mice via food intake in vivo, SD-169 is shown to prevent the development of type 1 diabetes in prediabetic mice and display therapeutic efficacy in treating animals already in mild and moderate hyperglycemic states.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Medicherla, S., et al. 2006. J. Pharmacol. Exp. Ther.318, 99.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service