Skip to Content
Merck
All Photos(1)

Key Documents

5.05767

Sigma-Aldrich

GSK-3beta Inhibitor XXVII, CESI

Synonym(s):

GSK-3beta Inhibitor XXVII, CESI, 2-(3-chloro-4-hydroxyphenyl)-8-(1,1-dioxidoisothiazolidin-2-yl)-3-hydroxy-6-methyl-4H-chromen-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€194.00

€194.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€194.00

About This Item

Empirical Formula (Hill Notation):
C19H16ClNO6S
CAS Number:
Molecular Weight:
421.85
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€194.00


Estimated to ship on17 May 2025


Request a Bulk Order

Assay

≥96% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

CC1=CC2=C(C(=C1)N3CCCS3(=O)=O)OC(=C(C2=O)O)C4=CC(=C(C=C4)O)Cl

General description

A cell-permeable, 3-hydroxychromone derived compound that acts as a selective and reversible inhibitor of GSK-3β (IC50<100 nM). Shown to increase phosphorylation at Ser9. Does not show any inhibitory effect on a panel of 15 other protein kinases, including AMPK and Akt. By blocking GSK-3β activity, it acts as a positive regulator of b-catenin signaling. Does not affect the viability or the growth rate of 3T3-L1 preadipocytes, however, it significantly reduces their differentiation into adipocytes (˜ 50 to 100 nM). Exerts anti-adipogenic activity by down-regulating C/EBPa and PPARg. Shown to reduce adipocyte size and diminish weight gain in diet-induced obese mice without affecting their daily food intake.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GSK3b

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

S. Lee et al. 2013, Biochemical Pharmacology85, 965.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service