Skip to Content
Merck
All Photos(1)

Key Documents

5.05277

Sigma-Aldrich

GTS-21 Dihydrochloride (DMBX-A)

Synonym(s):

GTS-21 Dihydrochloride (DMBX-A), E)-3-(2,4-dimethoxybenzylidene)-3,4,5,6-tetrahydro-2,3ʹ-bipyridine, DMBX-A, α7 nAChR Agonist, GTS-21, Alpha 7 Nicotinic Acetylcholine Receptor Agonist, GTS-21

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€112.00

€112.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€112.00

About This Item

Empirical Formula (Hill Notation):
C19H20N2O2 · 2HCl
CAS Number:
Molecular Weight:
381.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€112.00


Estimated to ship on23 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mM
H2O: 100 mM

storage temp.

−20°C

SMILES string

[Cl-].[Cl-].n1cc(ccc1)C2=NCCC\C\2=C/c3c(cc(cc3)OC)OC.[H+].[H+]

InChI

1S/C19H20N2O2.2ClH/c1-22-17-8-7-14(18(12-17)23-2)11-15-5-4-10-21-19(15)16-6-3-9-20-13-16;;/h3,6-9,11-13H,4-5,10H2,1-2H3;2*1H/b15-11+;;

InChI key

BXKYFUGAAFLYJL-BXGYHSFXSA-N

General description

A partial agonist selective for α7 nAChRs (EC50 = 10 µM). Used in neuroimmunology studies, cardiovascular disease research, and Alzheimer′s disease studies.

Biochem/physiol Actions

Primary Target
α7 nAChR
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kox, M., et al. 2011. Br. J. Anaesth.107, 559.
Kox, M., et al., 2011. Shock.36, 5.
Rosas-Ballina, M., et al. 2009. Mol. Med.15, 195.
Briggs, A., et al. 1997. Pharmacol. Biochem. Behav.57, 231.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service