Skip to Content
Merck
All Photos(1)

Key Documents

5.04539

Sigma-Aldrich

Eliprodil

Synonym(s):

Eliprodil, α-(4-Chlorophenyl)-4-[(4-fluorophenyl)methyl]-1-piperidineethanol, NMDA Antagonist IX

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€75.20

€75.20


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€75.20

About This Item

Empirical Formula (Hill Notation):
C20H23ClFNO
CAS Number:
Molecular Weight:
347.85
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€75.20


Estimated to ship on31 May 2025


Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 17 mg/mL

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)CC2CCN(CC2)CC(O)c3ccc(cc3)Cl

InChI

1S/C20H23ClFNO/c21-18-5-3-17(4-6-18)20(24)14-23-11-9-16(10-12-23)13-15-1-7-19(22)8-2-15/h1-8,16,20,24H,9-14H2

InChI key

GGUSQTSTQSHJAH-UHFFFAOYSA-N

General description

A negative allosteric modulator selective for NR2B containing NMDA receptors (IC50 = 1, >100, and >100 µM for NR2B, NR2A, and NR2C, respectively). Also a potent ligand for ς1 receptor (Ki = 0.013 µM), one of the two types of unique non-opioid, non-phencyclidine brain ς receptors. Blocks neuronal voltage-gated Ca2+ channels and selectively inhibits the rapid component of the delayed rectifier K+ current (IKr). A neuroprotective agent in acute ischemic stroke. Prolongs the QT interval in EKG.

Biochem/physiol Actions

Primary Target
NMDA receptors
Target IC50: 1, >100, and >100 →M for NR2B, NR2A, and NR2C, respectively.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lengyel, C., et al. 2004. Br. J. Pharmacol.143, 152.

Nikam, S., et al. 2002. Curr Pharm Des.8, 845.Whittemore, E., et al. 1997. J. Pharmacol. Exp. Ther.282, 326.M
Bath, C., et al. 1996. Eur. J. Pharmacol.299, 103.
Carter, C., et al. 1994. Neuropsychopharmacol.11, 257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service