Skip to Content
Merck
All Photos(1)

Key Documents

5.04519

Sigma-Aldrich

CIQ

Synonym(s):

CIQ, (3-Chlorophenyl) [3,4-dihydro-6,7-dimethoxy-1-[(4-methoxyphenoxy)methyl]-2(1H)-isoquinolinyl]methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26ClNO5
CAS Number:
Molecular Weight:
467.94
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

InChI

1S/C26H26ClNO5/c1-30-20-7-9-21(10-8-20)33-16-23-22-15-25(32-3)24(31-2)14-17(22)11-12-28(23)26(29)18-5-4-6-19(27)13-18/h4-10,13-15,23H,11-12,16H2,1-3H3

InChI key

VYMILMYEENZHAR-UHFFFAOYSA-N

General description

A positive allosteric modulator selective for NR2C/D containing NMDA receptors (EC50 = 2.7 and 2.8 µM for NR2C and NR2D, respectively). Increases channel opening frequency of recombinant NR2C/D containing NMDA receptors by two-fold without altering mean open time or EC50 values for glutamate or glycine bindings. No effect on NR2A or NR2B subtypes.

Biochem/physiol Actions

Primary Target
NMDA receptors

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming is required for complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Mullasseril, P., et al. 2010. Nat. Commun.1, 1038.
Ogden, K., et al. 2013. Mol. Pharmacol.83, 1045.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service