Skip to Content
Merck
All Photos(1)

Documents

5.04379

Sigma-Aldrich

HIF Inhibitor VII

Synonym(s):

HIF Inhibitor VII, 0X3, Hypoxia-Inducible Factor Inhibitor VI, N-(3-Chloro-5-fluorophenyl)-4-nitrobenzo[c][1,2,5]oxadiazol-5-amine, 0X3, N-(3-Chloro-5-fluorophenyl)-4-nitrobenzo[c][1,2,5]oxadiazol-5-amine, Hypoxia-Inducible Factor Inhibitor VI, 504379

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6ClFN4O3
CAS Number:
Molecular Weight:
308.65
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

InChI

1S/C12H6ClFN4O3/c13-6-3-7(14)5-8(4-6)15-10-2-1-9-11(17-21-16-9)12(10)18(19)20/h1-5,15H

InChI key

CDQUJZKBRAFWNG-UHFFFAOYSA-N

General description

A cell-permeable benzoxadiazolamine that prevents HIF-2α-HIF-β/ARNT heterodimeration by perturbing HIF-2α Per-ARNT-Sim/PAS-B domain surface β-sheet conformation via a 1:1 stoichiometric binding (KD = 81 nM), while exhibiting no affinity toward HIF-1αPAS-B domain. Shown to be metabolically stable and non-cytotoxic in cultures and selectively inhibit hypoxia-induced HIF-2α DNA binding and HIF-2α-dependent EPO mRNA upregulation (1 to10 µM for 6 to 12 h), but not HIF-1α DNA binding or HIF-1α-dependent PGK1 mRNA upregulation, in Hep3B cells. Unlike HIF Inhibitors I-VI, this compound does not affect the cellular protein or mRNA levels of HIF-1α & HIF-2α.
A cell-permeable, metabolically stable (t1/2 = 14 h in 786-0 cultures; no metabolization up to 24 h in medium alone), non-cytotoxic (up to 30 µM & 24 h in 786-0 and Hep3B cultures) benzoxadiazolamine that prevents HIF-2α-HIF-β/ARNT (aryl hydrocarbon receptor nuclear translocator) heterodimeration by perturbing HIF-2α Per-ARNT-Sim/PAS-B domain surface β-sheet conformation via a 1:1 stoichiometric binding to HIF-2α PAS-B internal cavity (KD = 81 nM), while exhibiting no affinity toward HIF-1α PAS-B domain (KD >>5 µM). Shown to inhibit nuclear HIF-2α-ARNT, but not HIF-1α-ARNT, association in hypoxic Hep3B cells in a dose-dependent manner (0.1 to 10 µM) and selectively decrease hypoxia-induced HIF-2α DNA binding and HIF-2α-dependent EPO mRNA upregulation (% inhibition/time/dose = 60%/6 h/1 µM, 90%/6 h/10 µM, 45%/12 h/1 µM, 93%/12 h/10 µM), displaying little effect toward HIF-1α DNA binding or HIF-1α-dependent PGK1 mRNA upregulation. Unlike HIF Inhibitors I-VI, this compound does not affect the cellular protein or mRNA levels of HIF-1α & HIF-2α.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HIF-2α
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Wu, D., et al. 2015. Nature in press.
Scheuermann, T.H., et al. 2013. Nat. Chem. Biol.9, 271.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service