Skip to Content
Merck
All Photos(1)

Key Documents

4.90046

Sigma-Aldrich

1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide

for synthesis

Synonym(s):

1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20F6N2O4S2
CAS Number:
Molecular Weight:
422.41
MDL number:
UNSPSC Code:
12352107

Quality Level

form

liquid

pH

5 (20 °C in H2O)

mp

-6 °C

density

1.40 g/cm3

InChI

1S/C9H20N.C2F6NO4S2/c1-3-4-7-10(2)8-5-6-9-10;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h3-9H2,1-2H3;/q+1;-1

InChI key

HSLXOARVFIWOQF-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service