475935
MT1-MMP, Catalytic Domain, Human, Recombinant, E. coli
Synonym(s):
MMP-14 Catalytic Domain
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥90% (SDS-PAGE)
Quality Level
form
liquid
specific activity
≥140 mU/mg protein
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
−70°C
General description
Note: 1 mU = 1 milliunit.
Recombinant, human pro-MT1-MMP expressed in E. coli and activated to yield the active catalytic domain (amino acids 89-265). Degrades fibrillar collagen Types I, II, and III, as well as other extracellular matrix proteins, and activates MMP-2 and MMP-13 resulting in enhanced proteolysis of the matrix for cell migration and proliferation.
Recombinant, human pro-MT1-MMP expressed in E. coli and activated to yield the active catalytic domain (amino acids 89-265). The catalytic domain is comprised of amino acid residues Tyr89 to Gly265 of mature human MT1-MMP. Hydrolyzes fibrillar collagens I, II, and III, as well as other extracellular matrix proteins, and activates MMP-2 and MMP-13, resulting in enhanced proteolysis of the matrix for cell migration and proliferation.
Packaging
Please refer to vial label for lot-specific concentration.
Warning
Toxicity: Standard Handling (A)
Physical form
In 150 mM NaCl, 50 mM Tris-HCl, 5 mM CaCl₂, pH 7.5.
Reconstitution
Following initial thaw, aliquot and freeze (-70°C).
Other Notes
Amano, T., et al. 2005. Cell Res.15, 150.
Knauper, V., et al. 1996. J. Biol. Chem. 271, 17124.
Pei, D., and Weiss, S.J. 1996. J. Biol. Chem. 271, 9135.
Sato, H., et al. 1994. Nature370, 61.
Knight, C.G., et al. 1992. REBS Lett.296, 263.
Knauper, V., et al. 1996. J. Biol. Chem. 271, 17124.
Pei, D., and Weiss, S.J. 1996. J. Biol. Chem. 271, 9135.
Sato, H., et al. 1994. Nature370, 61.
Knight, C.G., et al. 1992. REBS Lett.296, 263.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service