Skip to Content
Merck
All Photos(1)

Key Documents

454578

Sigma-Aldrich

Anti-Pan-NMDA NR1 Mouse mAb (R1JHL)

lyophilized, clone R1JHL, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

15 μG
€410.00

€410.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
15 μG
€410.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€410.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

R1JHL, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Mouse monoclonal antibody lyophilized from cell culture supernatant. Recognizes the ~115 kDa subunit of the NMDAR (N-methyl-D-aspartate receptor) protein.
Recognizes the ~115 kDa NMDA receptor NR1 subunit in rat cortex tissue extracts.
This Anti-Pan-NMDA NR1 Mouse mAb (R1JHL) is validated for use in Immunoblotting for the detection of Pan-NMDA NR1.

Immunogen

Rat
a recombinant fusion protein consisting of amino acids 1-564 of rat NMDA NR1

Application

Immunoblotting (1:300, colorimetric)

Immunoprecipitation (3 µg/200 µg lysate)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from cell culture supernatant.

Reconstitution

Reconstitute in 50 µl PBS (137 mM NaCl, 7.5 mM Na₂HPO₄, 2.7 mM KCl, 1.5 mM KH₂P0₄, pH 7.4)Following reconstitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions.

Analysis Note

Positive Control
Rat cortex tissue extracts

Other Notes

Does not cross-react with other NMDAR subunits. For immunoblotting, dilute in TBST containing 5% nonfat milk and 0.05% sodium azide. Antibody may be used up to 5 times in immunoblotting experiments without significant loss in signal strength. Store diluted antibody at 4°C between uses. Variables associated with assay conditions will dictate the proper working dilution.
Laurie, D.J., and Seeburg, P.H. 1994. J. Neurosci.14, 3180
Ishii, T., et al. 1993. J. Biol. Chem.268, 2836.
Monyer, H., et al. 1992. Science256, 1217.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service