Skip to Content
Merck
All Photos(1)

Key Documents

444247

Sigma-Aldrich

MMP Inhibitor II

The MMP Inhibitor II, also referenced under CAS 203915-59-7, controls the biological activity of MMP. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP Inhibitor II, N-Hydroxy-1,3-di-(4-methoxybenzenesulphonyl)-5,5-dimethyl-[1,3]-piperazine-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N3O8S2
CAS Number:
Molecular Weight:
513.58
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C21H27N3O8S2/c1-21(2)13-23(33(27,28)17-9-5-15(31-3)6-10-17)20(19(25)22-26)24(14-21)34(29,30)18-11-7-16(32-4)8-12-18/h5-12,20,26H,13-14H2,1-4H3,(H,22,25)

InChI key

MCSWSPNUKWMZHM-UHFFFAOYSA-N

General description

A potent, reversible, broad range inhibitor of matrix metalloproteinases. Inhibits MMP-1 (IC50 = 24 nM), MMP-3 (IC50 = 18.4 nM), MMP-7 (IC50 = 30 nM), and MMP-9 (IC50 = 2.7 nM).
A potent, reversible, broad-range inhibitor of matrix metalloproteinases. Inhibits MMP-1 (IC50 = 24 nM), MMP-3 (IC50 = 18.4 nM), MMP-7 (IC50 = 30 nM), and MMP-9 (IC50 = 2.7 nM).

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: yes
Target IC50: 24 nM, 18.4 nM, 30 nM, 2.7 nM, against MMP-1, MMP-3, MMp-7, and MMP-9, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Requires vigorous vortexing for 60 seconds to completely solubilize the product.

Other Notes

Pikul, S., et al. 1998. J. Med. Chem. 41, 3568.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service