Skip to Content
Merck
All Photos(2)

Key Documents

430-10

Millipore

Centriprep® Ultrafiltration Centrifugal Filters

Regenerated Cellulose (10 kDa MWCO), 15 mL sample volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101

material

Nylon cap
polycarbonate collector (for filtrate)
polycarbonate reservoir (for sample)
polypropylene seal
regenerated cellulose membrane

sterility

non-sterile

product line

Milliplex®

feature

holdup volume 0.5 mL

species reactivity

(Multi-Species)

manufacturer/tradename

Centriprep®

parameter

15 mL sample volume

technique(s)

multiplexing: suitable
protein extraction: suitable
protein purification: suitable

L

135 cm

diam.

2.81 cm

filtration area

2.84 cm2

pore size

10 kDa NMWCO

application(s)

sample preparation

shipped in

ambient

General description

Centriprep centrifugal filters are disposable ultrafiltration devices used for purifying, concentrating, and desalting biological samples in the 2–15 mL volume range. These complete, ready-to-use ultrafiltration devices are designed for operation in most centrifuges that can accommodate 50 mL centrifuge tubes. They are easy to use and offer a high flow rate. Centriprep devices consist of a sample container with a twist-lock cap, a filtrate collector containing a low-adsorptive, Ultracel regenerated cellulose membrane and an air-seal cap for sample isolation.

Features & Benefits:
•For use with samples containing high levels of solutes
•Unique inverse flow mode of operation with large deadstop
•Fast sample processing
•Available with Ultracel membranes: (3, 10, 30 and 50 kDa NMWL)
•Fits standard swinging-bucket rotor for 50 mL tubes

Applications:
Concentration and Purification of Particle-laden Solutions, Solutions with High Solute Concentration, Separation of Low MW Solutes from Fermentation Broths, Cell Culture Media and Lysates
Filter Code: YM-10
Filter Type: Ultrafiltration

Features and Benefits

Minimum Final Concentrate Volume: 700 µL

Physical form

Color Code: Green

Legal Information

CENTRIPREP is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samantha M Logan et al.
Frontiers in molecular biosciences, 7, 97-97 (2020-06-13)
The fatty acid composition of a pre-hibernation diet can influence the depth and duration of metabolic suppression achieved by hibernators. More specifically, a diet high in n-6 polyunsaturated fatty acids (PUFAs) relative to n-3 PUFAs is essential to maximize torpor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service