Skip to Content
Merck
All Photos(1)

Key Documents

422682

Sigma-Aldrich

K ir1.1 Inhibitor, VU591

The Kir1.1 Inhibitor, VU591 controls the biological activity of Kir1.1. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

K ir1.1 Inhibitor, VU591

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12N6O5
CAS Number:
Molecular Weight:
368.30
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light gray

solubility

DMSO: 100 mg/mL, clear, dark brown

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable, benzoimidazole, renal outer medullary potassium channel (ROMK) blocker that selectively inhibits inward rectifier potassium channel, Kir1.1 (IC50 = 240 nM), in HEK-293 cells. At 10 µM, it displays no effect on Kir2.1, Kir2.3, Kir4.1, and Kir7.1, with 150-fold selectivity over Kir6.2. In addition, it exhibits a modest off-target effect on the GABAA receptor (IC50 = 6.2 µM) among a panel of cardiac and central nervous system ion channels and receptors.
The Kir1.1 Inhibitor, VU591 controls the biological activity of Kir1.1. This small molecule/inhibitor is primarily used for Neuroscience applications.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bhave, G., et al. 2011. Mol Pharmacol.79, 42.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service