Skip to Content
Merck
All Photos(2)

Key Documents

420202

Sigma-Aldrich

JMJD Histone Demethylase Inhibitor III

The JMJD Histone Demethylase Inhibitor III controls the biological activity of JMJD Histone Demethylase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

JMJD Histone Demethylase Inhibitor III, JHDM Inhibitor III, Histone Lysine Demethylase Inhibitor X, ( E)-4-(Hydroxy(4-(4-((naphthalen-1-ylcarbamoyloxy)methyl)benzylamino)butyl)amino)-4-oxobut-2-enoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29N3O6
CAS Number:
Molecular Weight:
491.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=CC2=C1C=CC=C2)OCC3=CC=C(CNCCCCN(O)C(/C=C/C(O)=O)=O)C=C3

InChI key

FOJNYQXAAPCEPX-CCEZHUSRSA-N

General description

A substrate (methyl lysine) and cofactor (α-ketoglutarate) mimicking bivalent benzylamino-butylamino-N-hydroxybutenoic acid compound that acts as a selective in vitro inhibitor of Jumonji domain-containing histone demethylases (JHDMs; IC50 = 4.3, 3.4, 5.9, 10 and 43 µM for JMJD2A, JMJD2C, JMJD2E, PHF8 and JMJD3, respectively) over LSD1, asparaginyl and prolyl hydroxylases, and histone deacetylases (IC50 = 54, 83, 31, 22, 620, 680 and >800 µM for PHD1, PHD2, PHD3, FIH, LSD1 and HDACs, respectively). The cellular active JHDM Inhibitor, Methylstat is also available (Cat. No. 420203).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Luo, X., et al. 2011. J. Am. Chem. Soc.133, 9451.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuelai Luo et al.
Journal of the American Chemical Society, 133(24), 9451-9456 (2011-05-19)
Histone methylations are important chromatin marks that regulate gene expression, genomic stability, DNA repair, and genomic imprinting. Histone demethylases are the most recent family of histone-modifying enzymes discovered. Here, we report the characterization of a small-molecule inhibitor of Jumonji C

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service