Skip to Content
Merck
All Photos(1)

Key Documents

420136

Sigma-Aldrich

JNK Inhibitor IX

The JNK Inhibitor IX, also referenced under CAS 312917-14-9, controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JNK Inhibitor IX, N-(3-Cyano-4,5,6,7-tetrahydro-1-benzothien-2-yl)-1-naphthamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€205.00

€205.00


Estimated to ship on01 June 2025



Select a Size

Change View
5 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C20H16N2OS · xH2O
CAS Number:
Molecular Weight:
332.42 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€205.00


Estimated to ship on01 June 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c2c(c(c1NC(=O)c3c4c(ccc3)cccc4)C#N)CCCC2

InChI

1S/C20H16N2OS/c21-12-17-15-9-3-4-11-18(15)24-20(17)22-19(23)16-10-5-7-13-6-1-2-8-14(13)16/h1-2,5-8,10H,3-4,9,11H2,(H,22,23)

InChI key

WQGDQGAFSDMBLA-UHFFFAOYSA-N

General description

A thienylnaphthamide compound that acts as a potent and ATP binding site-targeting inhibitor of JNK2 and JNK3 (pIC50 = 6.5 and 6.7, respectively) with little or no activity against JNK1, p38α, and a panel of more than 30 other kinases (pIC50<5.0). In contrast, SP600125 offers little selectivity between JNK1 and JNK3 (pIC50 = 6.4 and 5.9, respectively) under the same assay conditions.

Biochem/physiol Actions

Cell permeable: no
Primary Target
JNK2 and JNK3
Product competes with ATP.
Reversible: no
pIC50 = 6.5 and 6.7, against JNK2 and JNK3, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Angell, R.M., et al. 2007. Bioorg. Med. Chem. Lett.17, 1296.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service