Skip to Content
Merck
All Photos(1)

Key Documents

376650

Sigma-Aldrich

D-erythro-Sphingosine, N-Hexanoyl-

Biologically active, cell-permeable, non-physiological ceramide analog.

Synonym(s):

D-erythro-Sphingosine, N-Hexanoyl-, C₆ Ceramide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€78.30

€78.30


Estimated to ship on22 May 2025



Select a Size

Change View
5 MG
€78.30

About This Item

Empirical Formula (Hill Notation):
C24H47NO3
CAS Number:
Molecular Weight:
397.63
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

€78.30


Estimated to ship on22 May 2025


Quality Level

Assay

≥98% (TLC)

form

waxy solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 25 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

N(C(C(O)\C=C\CCCCCCCCCCCCC)CO)C(=O)CCCCC

InChI

1S/C24H47NO3/c1-3-5-7-8-9-10-11-12-13-14-15-16-18-19-23(27)22(21-26)25-24(28)20-17-6-4-2/h18-19,22-23,26-27H,3-17,20-21H2,1-2H3,(H,25,28)/b19-18+

InChI key

NPRJSFWNFTXXQC-VHEBQXMUSA-N

General description

Biologically active, cell-permeable, non-physiological ceramide analog. Induces a dramatic arrest in the G0/G1 phase of the cell cycle. Activates MAP kinase. Activates protein phosphatase 2A (PP2A) at 10 nM. Inhibits diacylglycerol accumulation and phospholipase D (PLD) activation in fibroblasts. Induces apoptosis in MOLT-4 leukemia cells.
Biologically active, cell-permeable, nonphysiological ceramide analog. Stimulates cytosolic serine/threonine protein phosphatase in T9 cells and induces phosphorylation on Thr-669 in A-431 cells by stimulation of ceramide-activated protein kinase. Induces a dramatic arrest in the Go/G1 phase of the cell cycle. Activates MAP kinase. Activates protein phosphatase 2A (PP2A) at 10 nM. Inhibits diacylglycerol accumulation and phospholipase D activation in fibroblasts. Induces apoptosis in Molt-4 leukemia cells.

Biochem/physiol Actions

Cell permeable: yes
Effective concentration: 10 nM in activating protein phosphatase 2A (PP2A)
Primary Target
MAP kinase, protein phosphatase 2A (PP2A)
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Jayadev, S., et al. 1995. J. Biol. Chem. 270, 2047.
Venable, M.E., et al. 1994. J. Biol. Chem. 269, 26040.
Dobrowsky, R.T., et al. 1993. J. Biol. Chem. 268, 15523.
Raines, M.A., et al. 1993. J. Biol. Chem.268, 14572.
Dobrowsky, R.T., and Hannun, Y.A. 1992. J. Biol. Chem.267, 5048.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service