Skip to Content
Merck
All Photos(1)

Key Documents

374813

Sigma-Aldrich

Hemocyanin, Keyhole Limpet, Megathura crenulata, High Purity, Endotoxin-Free, Glycerol

Hemocyanin, Keyhole Limpet, Megathura crenulata, High Purity, Endotoxin Free, is a large, multi-subunit, O2-carrying, metalloprotein that is used as a carrier protein in the production of antibodies.

Synonym(s):

Hemocyanin, Keyhole Limpet, Megathura crenulata, High Purity, Endotoxin-Free, Glycerol, KLH

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€498.00

€498.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€498.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

€498.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (size exclusion chromatography)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

impurities

≤12 EU/mg Endotoxin
≥1.5 x 10⁻³ copper to protein ratio
≥4 mg/mL Protein

shipped in

wet ice

storage temp.

2-8°C

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 75 mM NaCl, 40 mM sodium phosphate, 50% glycerol.

Preparation Note

KLH is an extremely large protein complex that has a tendency to aggregate and precipitate from solutions. Use a buffer when diluting KLH and maintain the magnesium concentration at ≥10 mM. Store KLH solutions at 4°C. DO NOT freeze, vortex or agitate to dissolve KLH. Freezing or agitating KLH will cause protein aggregation and precipitation.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service