Skip to Content
Merck
All Photos(1)

Documents

341597

Sigma-Aldrich

FGF-II, Mouse, Recombinant, E. coli

Synonym(s):

mFGF-II, Fibroblast Growth Factor-II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.1 ng/μg Endotoxins (ng/μg FGF-2)

color

white

shipped in

wet ice

storage temp.

−20°C

General description

Full-length, recombinant, mouse basic fibroblast growth factor expressed in E. coli. A multi-functional factor with chemotactic activity for numerous cell types. It plays a role in the proliferation of neuronal precursors and in neuronal differentiation. A potent angiogenic factor both in vivo and in vitro. Activates phospholipase D in endothelial cells. Mouse FGFII shares 95% identity with human FGFII. Useful for studies of cell activation and signaling.

Biochem/physiol Actions

ED₅₀: ≤0.5 ng/ml as determined by the dose-dependent proliferation of BALB/3T3 cells.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C. Avoid freeze/thaw cycles of solutions.
Reconstitute to 100 µg/ml with sterile H₂O.

Other Notes

Coulier, F. et al. 1997. J. Mol Evol.44, 43.
Claus, P. et al., 2003. J. Biol. Chem.278, 749.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service