Skip to Content
Merck
All Photos(1)

Key Documents

328009

Sigma-Aldrich

ERK Inhibitor III

The ERK Inhibitor III, also referenced under CAS 331656-92-9, controls the biological activity of ERK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

ERK Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10N6O5
CAS Number:
Molecular Weight:
318.25
UNSPSC Code:
12352200

Quality Level

Assay

≥85% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable furanyl-nitroaminoguanidine ERK-binding (KD of ~13 µM) compound that acts as a substrate-, but not ATP-, site-targeting inhibitor of ERK. Selectively inhibits the EGF-stimulated cellular phosphorylation of ERK substrates Rsk-1 and Elk-1, but not that of ERK1/2 or the anisomycin-induced phosphorylation of the p38 substrate ATP-2 in HeLa cells.

Biochem/physiol Actions

Cell permeable: yes
KD = ~13 µM against ERK
Primary Target
ERK
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Chen, F., et al. 2006. Bioorg. Med. Chem. Lett.16, 6281.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service