Skip to Content
Merck
All Photos(1)

Documents

324707

Millipore

ELUGENT Detergent, 50% Solution

Non-ionic detergent composed of a mixture of alkyl glucosides.

Synonym(s):

ELUGENT Detergent, 50% Solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.25

Quality Level

form

viscous liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear brown to yellow

shipped in

ambient

storage temp.

15-25°C

General description

Non-ionic detergent composed of a mixture of alkyl glucosides. Like n-octyl-β-D-glucopyranoside and TRITON X-100 Detergent, it is useful for solubilization and purification of membrane-bound proteins. Relatively low cost alternative to n-octyl-β-D-glucopyranoside and n-octyl-β-D-maltoside in large-scale solubilizations. HLB number: 13.1.
Non-ionic detergent composed of a mixture of alkyl glucosides. Like n-octyl-β-D-glucopyranoside and TRITON X-100, it is useful for solubilization and purification of membrane-bound proteins. Relatively low cost alternative to n-octyl-β-D-glucopyranoside and n-octyl-β-D-maltoside in large scale solubilizations. Has an HLB of 13.1.

Warning

Toxicity: Irritant (B)

Physical form

50% aqueous solution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service