250225
Cytochalasin B, Dihydro-
Induces changes in cell morphology and motility with no effect on sugar transport.
Synonym(s):
Cytochalasin B, Dihydro-
Select a Size
€133.00
Estimated to ship on01 May 2025
Select a Size
About This Item
€133.00
Estimated to ship on01 May 2025
Recommended Products
Quality Level
description
Merck USA index - 14, 2790
Assay
≥98% (TLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
methanol: 10 mg/mL
ethanol: 2 mg/mL
DMSO: 5 mg/mL
shipped in
ambient
storage temp.
−20°C
SMILES string
N1[C@H]([C@H]3[C@@]4(OC(=O)CC[C@@H](CCC[C@H](C\C=C\[C@H]4[C@@H](C(=C)[C@H]3C)O)C)O)C1=O)Cc2ccccc2
InChI
1S/C29H39NO5/c1-18-9-7-13-22(31)15-16-25(32)35-29-23(14-8-10-18)27(33)20(3)19(2)26(29)24(30-28(29)34)17-21-11-5-4-6-12-21/h4-6,8,11-12,14,18-19,22-24,26-27,31,33H,3,7,9-10,13,15-17H2,1-2H3,(H,30,34)/b14-8+/t18-,19-,22-,23+,24+,26+,27-,29-/m1/s1
InChI key
WIULKAASLBZREV-RXPQEOCGSA-N
General description
Biochem/physiol Actions
Changes in cell morphology and motility
Warning
Reconstitution
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Carc. 2
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service