Skip to Content
Merck
All Photos(1)

Key Documents

236011

Sigma-Aldrich

COX-2 Inhibitor I

The COX-2 Inhibitor I, also referenced under CAS 416901-58-1, controls the biological activity of COX-2. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

COX-2 Inhibitor I, LM-1685, Methyl [5-methylsulfonyl-1-(4-chlorobenzyl)-1H-2-indolyl]carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16ClNO4S
CAS Number:
Molecular Weight:
377.84
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

methanol: 1 mg/mL
DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable potent and selective inhibitor of COX-2 from human monocytes (IC50 = 650 nM) and in whole blood (IC50 = 4.3 µM). Displays only very weak activity against COX-1 from human platelets (IC50 >10 µM) and in whole blood (IC50 >100 µM).
A cell-permeable potent, selective inhibitor of cyclooxygenase-2 (COX-2) (IC50 = 650 nM, human monocytes COX-2). Shown to specifically inhibit COX-2 in human whole blood (IC50 = 4.3 µM). Displays only weak inhibitory activity against COX-1 from human platelets (19% inhibition at 10 µM) and in whole blood (25% inhibition at 100 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
COX-2 from human monocytes
Product does not compete with ATP.
Reversible: no
Target IC50: 650 nM against COX-2 from human monocytes; 4.3 µM against COX-2 in whole blood

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Palomer, A., et al. 2002. J. Med. Chem.45, 1402.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheol Ho Heo et al.
Journal of functional biomaterials, 14(4) (2023-04-27)
Cyclooxygenase-2 (COX-2) is a biomolecule known to be overexpressed in inflammation. Therefore, it has been considered a diagnostically useful marker in numerous studies. In this study, we attempted to assess the correlation between COX-2 expression and the severity of intervertebral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service