Skip to Content
Merck
All Photos(1)

Key Documents

236002

Sigma-Aldrich

Anti-COX2, C-Terminal Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Cyclooxygenase-2, Anti-PGHS-2, Anti-Prostaglandin H Synthase-2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€446.00

€446.00


Estimated to ship on22 April 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€446.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€446.00


Estimated to ship on22 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

bovine, human, mouse, sheep, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTGS2(5743)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~72 kDa COX2 protein.
Recognizes COX2. Does not cross-react with COX1.
This Anti-COX2, C-Terminal Rabbit pAb is validated for use in Immunoblotting, ELISA for the detection of COX2, C-Terminal.

Immunogen

a synthetic peptide corresponding to a 17-amino acid sequence near the C-terminus of human COX2

Application

Immunoblotting (1-10 µg/ml, chemiluminescence)

ELISA (1:10,000-1:100,000)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Does not cross-react with COX1. Variables associated with assay conditions will dictate the optimal working dilution.
Hla, T., and Neilson, K. 1992. Proc. Natl. Acad. Sci. USA 89, 7384.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Freedom H Holland et al.
Neuroscience letters, 566, 131-136 (2014-02-26)
Early life stress exposure (ELS) yields risk for psychiatric disorders that might occur though a population-specific mechanism that impacts prefrontal cortical development. Sex differences in ELS effects are largely unknown and are also essential to understand social and cognitive development.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service