Skip to Content
Merck
All Photos(1)

Documents

234415

Sigma-Aldrich

C5a Receptor Antagonist, W-54011

The C5a Receptor Antagonist, W-54011, also referenced under CAS 405098-33-1, controls the biological activity of C5a Receptor.

Synonym(s):

C5a Receptor Antagonist, W-54011, N-((4-Dimethylaminophenyl)methyl)-N-(4-isopropylphenyl)-7-methoxy-1,2,3,4-tetrahydronaphthalen-1-carboxamide, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H36N2O2 · xHCl
CAS Number:
Molecular Weight:
456.62 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 50 mg/mL
methanol: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C30H36N2O2.ClH/c1-21(2)23-11-16-26(17-12-23)32(20-22-9-14-25(15-10-22)31(3)4)30(33)28-8-6-7-24-13-18-27(34-5)19-29(24)28;/h9-19,21,28H,6-8,20H2,1-5H3;1H

InChI key

UKBJWRMNGCDKNL-UHFFFAOYSA-N

General description

A potent, specific, and orally active C5a receptor antagonist. Shown to inhibit the binding of 125I-rhC5a to human neutrophils with a Ki of 2.2 nM, as well as C5a-induced intracellular Ca2+-mobilization, chemotaxis and reactive oxygen species (ROS) generation (IC50 = 3.1 nM, 2.7 nM and 1.6 nM, respectively) with no agonist activity up to 10 µM. Shown to prevent C5a-induced neutrophenia in gerbils (100 µg/kg i.v. or 3-30 mg/kg orally).
A tetrahydronaphthalenyl carboxamide compound that acts as a potent, specific and orally active C5a receptor antagonist. Shown to inhibit the binding of 125I-rhC5a to human neutrophils with a Ki of 2.2 nM, as well as C5a-induced intracellular Ca2+-mobilization, chemotaxis and reactive oxygen species (ROS) generation (IC50 = 3.1 nM, 2.7 nM and 1.6 nM, respectively) with no agonist activity up to 10 µM. Shown to prevent C5a-induced neutrophenia in gerbils (100 µg/kg i.v. or 3-30 mg/kg orally).

Biochem/physiol Actions

Cell permeable: no
Primary Target
C5a Receptor
Product does not compete with ATP.
Reversible: no
Target IC50: 3.1 nM, 2.7 nM and 1.6 nM against C5a-induced intracellular Ca2+-mobilization, chemotaxis and reactive oxygen species (ROS) generation, respectively
Target Ki: 2.2 nM against the binding of 125I-rhC5a to human neutrophils

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sumichika, H., et al. 2002. J. Biol. Chem.277, 49403.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatjana Reif et al.
Cell reports. Medicine, 2(6), 100317-100317 (2021-07-02)
Polymorphonuclear neutrophils (PMNs), the most abundant white blood cells, are recruited rapidly to sites of infection to exert potent anti-microbial activity. Information regarding their role in infection with human immunodeficiency virus (HIV) is limited. Here we report that addition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service