Skip to Content
Merck
All Photos(1)

Key Documents

234400-M

Sigma-Aldrich

Complement, Rabbit Serum

Complement, Rabbit Serum, is tested for suitability for HLA-ABC serology. Useful as an active source of complement for use with hybridoma antibodies, alloantisera, and related applications.

Synonym(s):

Rabbit serum

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€163.00

€163.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€163.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25
form:
lyophilized solid

€163.00


Estimated to ship on28 May 2025


Request a Bulk Order

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

storage temp.

2-8°C

General description

Complement, Rabbit Serum is a native, rabbit serum complement. Tested for suitability for HLA-ABC serology. Total lympholytic complement activity determined under conditions of NIH two-step cytotoxicity test that uses defined HLA isoantisera. Activity present at a titer of at least 1:4. No intrinsic cytotoxicity against human peripheral blood lymphocytes. Intended for micro and macro lymphocyte cytotoxicity testing with human or animal sera. Also useful as an active source of complement for use with hybridoma antibodies, alloantisera, and related applications in which a non-cytotoxic source is required.

Application

Complement, rabbit serum has been used:
  • in complement killing of fibroblasts for the isolation of Schwann cells (SCs)[1]
  • as a component of Dulbecco′s modified Eagle medium (DMEM) for the purification of SC cells[2]
  • to eliminate fibroblasts for the SC isolation[3]

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for 24 h at 4°C or for up to 1 month at -20°C.
Reconstitute with 5 ml of dH₂O.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Colombelli et al.
The Journal of cell biology, 208(3), 313-329 (2015-02-04)
Fast neural conduction requires accumulation of Na(+) channels at nodes of Ranvier. Dedicated adhesion molecules on myelinating cells and axons govern node organization. Among those, specific laminins and dystroglycan complexes contribute to Na(+) channel clustering at peripheral nodes by unknown
Xiaona Feng et al.
Communications biology, 3(1), 716-716 (2020-11-29)
Transient receptor potential vanilloid 4 (TRPV4) is a non-selective calcium-permeable cation channel that is widely expressed and activated in various neurons and glial cells in the nervous system. Schwann cells (SCs) are primary glia cells that wrap around axons to
Kathleen K Catignas et al.
Glia, 69(1), 91-108 (2020-08-04)
In the developing peripheral nervous system, Schwann cells (SCs) extend their processes to contact, sort, and myelinate axons. The mechanisms that contribute to the interaction between SCs and axons are just beginning to be elucidated. Using a SC-neuron coculture system

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service