Skip to Content
Merck
All Photos(2)

Documents

23-042-M

Sigma-Aldrich

Src Protein, 10 µg

Full length, recombinant human Src. N-terminal c-Myc, 6His-tagged. For use in Enzyme Assays. Functions as a substrate in ubiquitination assays.

Synonym(s):

Proto-oncogene c-Src, pp60c-src, SRC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

General description

Src is a ubiquitously expressed, non-receptor tyrosine kinase that plays a pivotal role in numerous cellular processes such as proliferation, migration, and transformation. Due to its critical function in regulating signal transduction pathways, Src activity is tightly controlled through reversible phosphorylation and dephosphorylation events and down-regulation by ubiquitin-dependent degradation. Dysfunction of Src, through overexpression or increased activation has profound effects on basic cellular functions. Elevated Src expression and/or activation is evident across a wide range of solid tumour types and has been strongly implicated in the development, maintenance, progression, and metastatic spread of several human cancers, such as prostate, lung, breast, and colorectal cancer.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service