Skip to Content
Merck
All Photos(2)

Key Documents

23-039-M

Sigma-Aldrich

Smad7

Synonym(s):

Mothers against decapentaplegic homologue 7, MAD homologue 7, Mothers against DPP homologue 7, Mothers against decapentaplegic homologue 8, MADH7, MADH8

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€682.00

€682.00


Estimated to ship on17 May 2025



Select a Size

Change View
10 μG
€682.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

€682.00


Estimated to ship on17 May 2025


biological source

human

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

General description

The TGF-β signalling cascade regulates diverse cellular processes, including cell proliferation, differentiation, apoptosis, adhesion and migration. It plays a fundamental role in a number of different physiological and pathological processes, such as embryonic development, adult homoeostasis, tumourigenesis and tissue fibrogenesis. Smad7 has been identified as a key negative regulator of TGF-β signalling, antagonising it through multiple mechanisms in both the cytoplasm and in the nucleus. Altered expression of Smad7 is often associated with human diseases, such as cancer, tissue fibrosis and inflammatory diseases.

Application

Ubiquitination Cascade Comoponent: Substrate

Physical form

Purified using immobilized metal affinity chromatography.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service