Skip to Content
Merck
All Photos(1)

Key Documents

220467

Sigma-Aldrich

2-Chloro-2′-deoxyadenosine

A deoxyadenosine analog that displays strong antileukemic and immunosuppressive activity.

Synonym(s):

2-Chloro-2′-deoxyadenosine, 2CdA, Cladribine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€83.60

€83.60


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€83.60

About This Item

Empirical Formula (Hill Notation):
C10H12ClN5O3
CAS Number:
Molecular Weight:
285.69
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€83.60


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 2337

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1nc2[n](cnc2c(n1)N)[C@@H]3O[C@@H]([C@H](C3)O)CO

InChI

1S/C10H12ClN5O3/c11-10-14-8(12)7-9(15-10)16(3-13-7)6-1-4(18)5(2-17)19-6/h3-6,17-18H,1-2H2,(H2,12,14,15)/t4-,5+,6+/m0/s1

InChI key

PTOAARAWEBMLNO-KVQBGUIXSA-N

General description

A deoxyadenosine analog that displays strong antileukemic and immunosuppressive activity. Also induces apoptosis in chronic lymphocytic leukemia (CLL) cells.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Antileukemic and immunosuppressive activity
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Other Notes

Bellosillo, B., et al. 1997. Blood 89, 3378; Bryson, H.M., and Sorkin, E.M. 1993. Drugs 46, 872.
Robertson, L.E., et al. 1993. Blood81, 143.
Carson, D.A., et al. 1988. Adv. Enzyme Regul. 27, 395.
Carson, D.A., et al. 1984. Proc. Natl. Acad. Sci. USA 81, 2232.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service