Skip to Content
Merck
All Photos(1)

Key Documents

219358

Sigma-Aldrich

Anti-Cathepsin G Rabbit pAb

Sign Into View Organizational & Contract Pricing


About This Item

antibody form

purified antibody

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

isotype

IgG

General description

Rabbit polyclonal antibody purified by ammonium sulfate precipition. Recognizes the cathepsin G protein.
Recognizes cathepsin G.

Immunogen

purified human neutrophil cathepsin G

Application

ELISA (≥1:1000)
Immunoblotting (see application references)
Immunoelectrophoresis (see comments)

Physical form

In PBS, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Monospecific for cathepsin G as determined by a single arc upon immunoelectrophoresis against cathepsin G. Variables associated with assay conditions will dictate the proper working dilution.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service