Skip to Content
Merck
All Photos(1)

Documents

203637-M

Millipore

Anti-Bone Sialoprotein Mouse mAb (ID1.2)

lyophilized, clone ID1.2, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

ID1.2, monoclonal

form

lyophilized

contains

≤0.1% sodium azide as preservative
0.04% Thimerosal as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Anti-Bone Sialoprotein, mouse monoclonal, clone ID1.2, recognizes ~70-80 kDa bone sialoprotein in osteoblasts and osteocytes. It is validated for ELISA, Western blotting, and for paraffin sections.
Purified mouse monoclonal antibody. Recognizes the ~70-80 kDa bone sialoprotein.
Recognizes the ~70-80 kDa bone sialoprotein in osteoblasts and osteocytes.

Immunogen

Human
purified human bone sialoprotein

Application

ELISA (see comments)

Immunoblotting (1:1000)

Paraffin Sections (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 50 mM Tris, pH 7.4.

Reconstitution

Reconstitute with dH₂O. Please see label for lot-specific reconstitution volume.

Analysis Note

Positive Control
Human osteoblasts or osteocytes

Other Notes

Does not cross-react with human osteocalcin, osteonectin, osteopontin, or bone alkaline phosphatase. This antibody has also been reported to work for ELISA and paraffin sections. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service