Skip to Content
Merck
All Photos(1)

Key Documents

194933

Sigma-Aldrich

AZOCOLL Substrate, >100 Mesh

solid, proteolytic substrate, Calbiochem®

Synonym(s):

AZOCOLL Substrate, >100 Mesh

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€117.00
5 G
€643.00

€117.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€117.00
5 G
€643.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€117.00


Estimated to ship on28 May 2025


Request a Bulk Order

Product Name

AZOCOLL Substrate, >100 Mesh, General proteolytic substrate.

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark red

solubility

PBS: 10 mg/mL

shipped in

ambient

storage temp.

10-30°C

General description

General proteolytic substrate. Smaller particle size provides a rapid, simple method for quantitative determination of protease content in aqueous solutions. Color yield (520 nm): >1.0; blank (520 nm, 50 mg/5 ml buffer, 10 minutes at 37°C, pH 7.0): <0.3. Mesh: >100.

Biochem/physiol Actions

Cell permeable: no
Primary Target
General proteolytic substrate
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Poilane, I., et al. 1998. Can. J. Microbiol. 44, 157.
Braganza, V.J., and Simmons, W.H. 1991. Biochemistry 30, 4997.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service