Skip to Content
Merck
All Photos(1)

Key Documents

19-203

Sigma-Aldrich

Epoxomicin Inhibitor

The Epoxomicin Inhibitor controls the biological activity of Epoxomicin. This small molecule/inhibitor is primarily used for Biochemicals applications.

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€366.00

€366.00


Estimated to ship on04 May 2025



Select a Size

Change View
100 μG
€366.00

About This Item

UNSPSC Code:
12352106
eCl@ss:
32160405
NACRES:
NA.41

€366.00


Estimated to ship on04 May 2025


Quality Level

form

solid

manufacturer/tradename

Upstate®

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Application

Cell permeable, potent and specific irreversible inhibitor of the proteasome. Novel mechanism of action. Blocks chymotrypsin-like, trypsin like and PGPH activities of the proteasome. Regulates antigen presentation at non-toxic doses. Displays in vivo anti-inflammatory activity in mouse ear inflammation assay and in vitro NF-kB inhibition. The product is not sterile.

Biochem/physiol Actions

Inhibitor Type: Protease Inhibitors
Protein Target: RPS27A

Storage and Stability

Store at -20°C for up to 1 year. Store solutions at -20°C for up to 3 months.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Schwarz et al.
Journal of immunology (Baltimore, Md. : 1950), 164(12), 6147-6157 (2000-06-08)
The complete inhibition of proteasome activities interferes with the production of most MHC class I peptide ligands as well as with cellular proliferation and survival. In this study we have investigated how partial and selective inhibition of the chymotrypsin-like activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service