1.50619
SAMS Installation Kit
tubing L 5 m, pkg of 5 ea (fittings), for use with CARS system (SEQUANT™)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pkg of 5 ea (fittings)
Quality Level
technique(s)
ion chromatography: suitable
tubing L
5 m
compatibility
for use with CARS system (SEQUANT™)
General description
SeQuant™ CARS (Continuous Anion Regeneration System) with SAMS (SeQuant Anion Membrane Suppressor) will enhance and simplify your anion chromatography analysis. The continuous chemical suppression of the eluent reduces the background and increases the signal for the analyte anions.
This tubing kit is required for the installation of SAMS and is also included in the CARS system.
This tubing kit is required for the installation of SAMS and is also included in the CARS system.
tubing (5 m) and fittings (6 ea) for CARS system SeQuant
Analysis Note
Appearance: conforms
Legal Information
SeQuant is a trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service