Skip to Content
Merck
All Photos(1)

Key Documents

1.50616

Supelco

ULB-P Regeneration Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

technique(s)

ion chromatography: suitable

Quality Level

pH

2 (20 °C in H2O)

bp

100 °C/1013 hPa

density

1 g/cm3 at 20 °C

storage temp.

no temp limit

General description

SeQuant CARS (Continuous Anion Regeneration System) with SAMS (SeQuant Anion Membrane Suppressor) will enhance and simplify your anion chromatography analysis. The continuous chemical suppression of the eluent reduces the background and increases the signal for the analyte anions.

This is a refill for the ULB-P supplied with the CARS system. New ULB-P regeneration solution is required when the cartridge is exchanged.

Analysis Note

Assay: 72.0 - 88.0 mmol/l
Appearance (clearness): conforms
Appearance (colour): conforms

Legal Information

SeQuant is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service