1.50613
CARS Cartridge Small
suitable for ion chromatography, for use with SEQUANT™ System
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
technique(s)
ion chromatography: suitable (high capacity)
Quality Level
compatibility
for use with SEQUANT™ System
General description
SeQuant™ CARS (Continuous Anion Regeneration System) with SAMS (SeQuant Anion Membrane Suppressor) will enhance and simplify your anion chromatography analysis. The continuous chemical suppression of the eluent reduces the background and increases the signal for the analyte anions.
This cartridge replaces the one supplied with the CARS system when it is depleted.
This cartridge replaces the one supplied with the CARS system when it is depleted.
high-capacity ion-exchanger (0.5L, 0.9eq) for CARS system SeQuant
Analysis Note
Appearance: conforms
Legal Information
SeQuant is a trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service