1.50611
CARS Complete System (230V,50/60Hz)
for use with SeQuant™ anion membrane supressor
All Photos(1)
About This Item
technique(s)
ion chromatography: suitable
Quality Level
compatibility
for use with SeQuant™ anion membrane supressor
General description
SeQuant™ CARS (Continuous Anion Regeneration System) with SAMS (SeQuant Anion Membrane Suppressor) will enhance and simplify your anion chromatography analysis. The continuous chemical suppression of the eluent reduces the background and increases the signal for the analyte anions.
This CARS system requires a SAMS suppressor for operation.
This CARS system requires a SAMS suppressor for operation.
continuous auto-regeneration system for SAMS suppressors SeQuant
Legal Information
SeQuant is a trademark of Merck KGaA, Darmstadt, Germany