Skip to Content
Merck
All Photos(1)

Key Documents

14-910-M

Sigma-Aldrich

PI3K-C2γ Protein, 10 µg

N-terminal GST-tagged recombinant human PI3K-C2γ, amino acids 468–1203, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€985.00

€985.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€985.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€985.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 112 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

GenBank accession no.

UniProt accession no.

Gene Information

human ... PIK3C2G(5288)

General description

N-terminal GST-tagged recombinant human PI3K-C2γ amino acids 468–1203
Product Source: Expressed by baculovirus in Sf21 insect cells

Application

Research Category
Signaling
Research Sub Category
Inflammation & Autoimmune Mechanisms

Oncology

Diabetes

Biochem/physiol Actions

Protein Target: PI3K-C2γ
Target Sub-Family: PI3K/PI4K

Packaging

Also available in 250μg size as catalog # 14-910M - Call for pricing and availability.

Quality

Routinely evaluated in the PIProfiler.

Physical form

0.54mg/ml of enzyme in 50mM Tris/HCl pH7.5, 300mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 0.2mM PMSF, 1mM benzamidine, 0.1% 2-mercaptoethanol. Frozen solution.
Glutathione agarose affinity chromatography

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service