1.46698
Sodium Chloride Peptone Buffer
ready-to-use, bottle volume 500 mL , filling volume
Synonym(s):
NaCl Peptone Buffer, Peptone Buffer, Sodium Chloride Buffer
About This Item
Recommended Products
Agency
EP 2.6.13
JP 5.02
USP 62
Quality Level
description
with 1% Tween® 80
form
liquid
feature
closure type screw cap with septum
ready-to-use
packaging
pack of 6 × 500 mL each
bottle of
technique(s)
direct inoculation: suitable
sterile filtration: suitable
bottle capacity
500 mL
bottle volume
500 mL , filling volume
pH range
6.8-7.2
application(s)
bioburden testing
pharmaceutical
components
tween
storage temp.
2-25°C
suitability
nonselective for
General description
The neutral pH range is optimum for the viability of microorganisms′ present. The medium aims at maximum recovery of microorganisms present in the test sample.
The addition of Tween® 80(polysorbate) assists the suspension of hydrophobic substances and helps neutralize disinfectant agents like quaternary ammonium compounds, iodine, and parabens.
Application
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service